missing translation for 'onlineSavingsMsg'
Learn More

Potassium Biiodate, Certified, 0.0375N ±0.0005N (0.00313M), LabChem™

Catalog No. LC187372
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC187372 1 L
LC187371 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC187372 Supplier LabChem Supplier No. LC187372

Chemical Identifiers

CAS 13455-24-8 , 7732-18-5
Molecular Formula HI2KO6
Molecular Weight (g/mol) 389.909
InChI Key ACAYDTMSDROWHW-UHFFFAOYSA-M
Synonym potassium hydrogen diiodate, potassium biiodate, kaliumhydrogendijodat, iodic acid hio3 , potassium salt 2:1, potassium biiodate solution, potassium iodic acid iodate, io3.k.hio3, potassium ion iodic acid iodate, potassium hydrogen iodate powder, potassium hydrogen diiodate, p.a
PubChem CID 23700942
IUPAC Name potassium;iodic acid;iodate
SMILES OI(=O)=O.[O-]I(=O)=O.[K+]

Specifications

Color Colorless
CAS Max % 99.88, 0.12
Physical Form Liquid
Quantity 1 L
Linear Formula KHI2O6
Solubility Information Soluble in water
Formula Weight 389.92
Grade Certified
Concentration or Composition (by Analyte or Components) 0.0375N ±0.0005N (0.00313M)
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Decomposition Information Iodine vapor
Density 1g/mL
Chemical Name or Material Potassium Biiodate
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature