missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Biiodate, 0.100 N (N/10), Ricca Chemical
EPA for Chlorine Testing
Chemical Identifiers
| CAS | 13455-24-8 , 7732-18-5 |
|---|---|
| Molecular Formula | HI2KO6 |
| Molecular Weight (g/mol) | 389.909 |
| InChI Key | ACAYDTMSDROWHW-UHFFFAOYSA-M |
| Synonym | potassium hydrogen diiodate, potassium biiodate, kaliumhydrogendijodat, iodic acid hio3 , potassium salt 2:1, potassium biiodate solution, potassium iodic acid iodate, io3.k.hio3, potassium ion iodic acid iodate, potassium hydrogen iodate powder, potassium hydrogen diiodate, p.a |
| PubChem CID | 23700942 |
| IUPAC Name | potassium;iodic acid;iodate |
| SMILES | OI(=O)=O.[O-]I(=O)=O.[K+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
587916
|
N/A | 500 mL | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
587932
|
N/A | 1 L | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 13455-24-8 , 7732-18-5 | |
| 389.909 | |
| potassium hydrogen diiodate, potassium biiodate, kaliumhydrogendijodat, iodic acid hio3 , potassium salt 2:1, potassium biiodate solution, potassium iodic acid iodate, io3.k.hio3, potassium ion iodic acid iodate, potassium hydrogen iodate powder, potassium hydrogen diiodate, p.a | |
| potassium;iodic acid;iodate |
| HI2KO6 | |
| ACAYDTMSDROWHW-UHFFFAOYSA-M | |
| 23700942 | |
| OI(=O)=O.[O-]I(=O)=O.[K+] |
Specifications
| 13455-24-8 , 7732-18-5 | |
| 100.0, 0.33 | |
| potassium hydrogen diiodate, potassium biiodate, kaliumhydrogendijodat, iodic acid hio3 , potassium salt 2:1, potassium biiodate solution, potassium iodic acid iodate, io3.k.hio3, potassium ion iodic acid iodate, potassium hydrogen iodate powder, potassium hydrogen diiodate, p.a | |
| OI(=O)=O.[O-]I(=O)=O.[K+] | |
| 389.909 | |
| 0.100Normal |
| 98.0, 0.32 | |
| HI2KO6 | |
| ACAYDTMSDROWHW-UHFFFAOYSA-M | |
| potassium;iodic acid;iodate | |
| 23700942 | |
| Potassium Biiodate |