missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bifluoride (Potassium Hydrogen Fluoride), Reagents

Catalog No. 01337123
Change view
Click to view available options
Quantity: 2.5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01337123 Quantity: 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01337123 Supplier Labchem Inc Supplier No. C22771002.5A5

Potassium Bifluoride (Potassium Hydrogen Fluoride), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7789-29-9
Molecular Formula F2HK
Molecular Weight (g/mol) 78.10
InChI Key VBKNTGMWIPUCRF-UHFFFAOYSA-M
Synonym Potassium Hydrogen Fluoride, Potassium Fluoride Hydrofluoride
IUPAC Name potassium hydrogen difluoride
SMILES [H+].[F-].[F-].[K+]

Specifications

Melting Point 239°C
Color Colorless
Physical Form Solid
Quantity 2.5 kg
Grade Reagent
Packaging Poly Bottle
Specific Gravity 2.3743
Chemical Name or Material Potassium Bifluoride (Potassium Hydrogen Fluoride)