missing translation for 'onlineSavingsMsg'
Learn More

Potassium Amylxanthate 97.0+%, TCI America™

Catalog No. A046125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A046125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A046125G Supplier TCI America Supplier No. A046125G
Only null left

Chemical Identifiers

CAS 2720-73-2
Molecular Formula C6H11KOS2
Molecular Weight (g/mol) 202.37
MDL Number MFCD00059150
InChI Key YIBBMDDEXKBIAM-UHFFFAOYSA-M
Synonym Amylxanthic Acid Potassium Salt, Potassium O-Amyl Dithiocarbonate
PubChem CID 493949
IUPAC Name potassium [(pentyloxy)methanethioyl]sulfanide
SMILES [K+].CCCCCOC([S-])=S

Specifications

Quantity 25 g
Formula Weight 202.37
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium Amylxanthate
EINECSNumber (2)-1787
RTECSNumber FG1581900
TSCA Yes