accessibility menu, dialog, popup

UserName

KH3COSS

CAS RN 2720-73-2

IUPAC Name: potassium [(pentyloxy)methanethioyl]sulfanide
Synonyms: potassium [(pentyloxy)methanethioyl]sulfanide
Molecular Weight (g/mol): 202.37
Molecular Formula: C6H11KOS2
InChi Key: YIBBMDDEXKBIAM-UHFFFAOYSA-M
SMILES: [K+].CCCCCOC([S-])=S
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results