missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Allyltrifluoroborate 97.0+%, TCI America™
$119.30 - $585.32
Chemical Identifiers
| CAS | 233664-53-4 |
|---|---|
| Molecular Formula | C3H5BF3K |
| Molecular Weight (g/mol) | 147.977 |
| InChI Key | TVPZAMJXLCDMIT-UHFFFAOYSA-N |
| PubChem CID | 23664277 |
| IUPAC Name | potassium;trifluoro(prop-2-enyl)boranuide |
| SMILES | [B-](CC=C)(F)(F)F.[K+] |
Chemical Identifiers
| 233664-53-4 | |
| 147.977 | |
| 23664277 | |
| [B-](CC=C)(F)(F)F.[K+] |
| C3H5BF3K | |
| TVPZAMJXLCDMIT-UHFFFAOYSA-N | |
| potassium;trifluoro(prop-2-enyl)boranuide |
Specifications
| 233664-53-4 | |
| C3H5BF3K | |
| TVPZAMJXLCDMIT-UHFFFAOYSA-N | |
| potassium;trifluoro(prop-2-enyl)boranuide | |
| 23664277 | |
| ≥97.0% (W) | |
| Potassium Allyltrifluoroborate |
| White | |
| 1 g | |
| [B-](CC=C)(F)(F)F.[K+] | |
| 147.977 | |
| 147.98 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes