accessibility menu, dialog, popup

UserName

BCH2FFFK

CAS RN 233664-53-4

IUPAC Name: potassium trifluoro(prop-2-en-1-yl)boranuide
Synonyms: potassium trifluoro(prop-2-en-1-yl)boranuide
Molecular Weight (g/mol): 147.98
Molecular Formula: C3H5BF3K
InChi Key: TVPZAMJXLCDMIT-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)CC=C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results