missing translation for 'onlineSavingsMsg'
Learn More

Potassium Acid Phthalate, Certified, 0.0500N ±0.0005N (0.05M), LabChem™

Catalog No. LC187001
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC187001 500 mL Poly Bottle
LC187002 1 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC187001 Supplier LabChem Supplier No. LC187001

Chemical Identifiers

CAS 877-24-7 , 7732-18-5
Molecular Formula C8H5KO4
Molecular Weight (g/mol) 204.222
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
Synonym potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic
PubChem CID 23676735
IUPAC Name potassium;2-carboxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]

Specifications

CAS Max % 98.98, 1.02
Decomposition Information Carbon monoxide; Carbon dioxide
Color Colorless
Density 1g/mL
Packaging Poly Bottle
Linear Formula KHC8H4O4
Quantity 500 mL
Specific Gravity 1g/mL
Solubility Information Soluble in water
Formula Weight 204.23
Grade Certified
Concentration or Composition (by Analyte or Components) 0.0500N ±0.0005N (0.05M)
Physical Form Liquid
Chemical Name or Material Potassium Acid Phthalate
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature