missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Acid Phthalate, ACS Reagent, Primary Standard (Potassium Hydrogen Phthalate), Reagents
Potassium Acid Phthalate, ACS Reagent, Primary Standard (Potassium Hydrogen Phthalate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C1276500250A6
Specifications
| Potassium Acid Phthalate, ACS Reagent, Primary Standard (Potassium Hydrogen Phthalate) | |
| 877-24-7 | |
| White | |
| Potassium Hydrogen Phthalate, KHP, Potassium Biphthalate | |
| ACS | |
| Poly Bottle | |
| 1.636 |
| 0°C | |
| 378°C | |
| HOCOC6H4COOK | |
| 204.22 | |
| 250 g | |
| Solid |