missing translation for 'onlineSavingsMsg'
Learn More

Potassium Acid Phthalate, ACS Reagent, Primary Standard (Potassium Hydrogen Phthalate), Reagents

Catalog No. p-200021009
Change view
Click to view available options
Quantity:
125 g
250 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337876 125 g
01337875 250 g
01337874 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337876 Supplier Labchem Inc Supplier No. C1276500125A6
Only null left

Potassium Acid Phthalate, ACS Reagent, Primary Standard (Potassium Hydrogen Phthalate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Potassium Acid Phthalate, ACS Reagent, Primary Standard (Potassium Hydrogen Phthalate)
CAS 877-24-7
Melting Point 0°C
Boiling Point 378°C
Color White
Molecular Formula HOCOC6H4COOK
Synonym Potassium Hydrogen Phthalate, KHP, Potassium Biphthalate
Molecular Weight (g/mol) 204.22
Grade ACS
Quantity 125 g
Packaging Poly Bottle
Physical Form Solid
Specific Gravity 1.636
Show More Show Less