missing translation for 'onlineSavingsMsg'
Learn More

Potassium Acid Phthalate, 300 mg/L, Multi-Purpose Standard, Ricca Chemical

Catalog No. 5867316
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
5867316 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5867316 Supplier Ricca Chemical Company Supplier No. 5867.316
Only null left

Chemical Identifiers

CAS 7732-18-5 , 877-24-7
Molecular Formula C8H5KO4
Molecular Weight (g/mol) 204.222
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
Synonym potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic
PubChem CID 23676735
IUPAC Name potassium;2-carboxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]

Specifications

Color Colorless
CAS Min % 98.0, 0.03
CAS Max % 100.0, 0.03
Physical Form Liquid
Quantity 500 mL
Grade Laboratory
Concentration or Composition (by Analyte or Components) 300mg/L, Multi-Purpose Standard
Packaging Natural Poly Bottle
Chemical Name or Material Potassium Acid Phthalate
ShelfLife 12 months