missing translation for 'onlineSavingsMsg'
Learn More

Potassium Acetate, puriss., Honeywell Fluka™

Catalog No. p-7071941
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002225 1 kg
6002227 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6002225 Supplier Honeywell Chemicals Supplier No. 250591KG
Only null left

Puriss., meets analytical specification of BP, Ph. Eur., E261, 99-101%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 127-08-2
Molecular Formula C2H3KO2
Molecular Weight (g/mol) 98.142
MDL Number MFCD00012458
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
Synonym potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
PubChem CID 517044
ChEBI CHEBI:32029
IUPAC Name potassium;acetate
SMILES CC(=O)[O-].[K+]

Specifications

Quantity 1 kg
Linear Formula CH3COOK
UN Number NONH for all modes of transport
Beilstein 3595449
Formula Weight 98.14g/mol
Percent Purity 99 to 101%
Grade puriss.
Density 1.57g/cm3at 25°C
Chemical Name or Material Potassium Acetate, Meets analytical specification of Ph. Eur. BP
EINECSNumber 204-822-2
RTECSNumber AJ3325000