missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Potassium Acetate, Molecular Biology Grade, Calbiochem™,

Catalog No. p-5547642
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
529543250GM 250 g
5295431KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 529543250GM Supplier MilliporeSigma™ Supplier No. 529543250GM
Only null left
Hygroscopic

Chemical Identifiers

CAS 127-08-2
Molecular Formula C2H3KO2
Molecular Weight (g/mol) 98.142
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
Synonym potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
PubChem CID 517044
ChEBI CHEBI:32029
IUPAC Name potassium;acetate
SMILES CC(=O)[O-].[K+]

Specifications

Quantity 250 g
Assay Percent Range ≥99 to 100%
Solubility Information Easily soluble in cold water
Formula Weight 98.1
Chemical Name or Material Potassium Acetate
Grade by perchloric acid titration (dry basis)
EINECSNumber 204-822-2
RTECSNumber AJ3325000
Recommended Storage Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).