Learn More
Potassium acetate, 99%
CAS: 127-08-2 | C2H3KO2 | 98.142 g/mol
Supplier: Thermo Scientific Chemicals A1632136
Description
Potassium acetate is used as deicer to remove snow and ice from the surface and considered as a replacement for calcium chloride or magnesium chloride. It is used as a food additive, as a buffer in biological research, as an extinguishing agent used in class K fire extinguishers and as a catalyst to produce polyurethane polymers. It is utilized for the precipitation of dodecyl sulfate bound proteins, thereby removal of proteins from DNA. It plays an important role in preserving tissues, fixation and mummification.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPotassium acetate is used as deicer to remove snow and ice from the surface and considered as a replacement for calcium chloride or magnesium chloride. It is used as a food additive, as a buffer in biological research, as an extinguishing agent used in class K fire extinguishers and as a catalyst to produce polyurethane polymers. It is utilized for the precipitation of dodecyl sulfate bound proteins, thereby removal of proteins from DNA. It plays an important role in preserving tissues, fixation and mummification.
Solubility
Soluble in water, alcohol and ammonia. Insoluble in acetone and ether.
Notes
Hygroscopic. Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications
| Potassium acetate | |
| 292°C | |
| C2H3KO2 | |
| 500 g | |
| Hygroscopic | |
| potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 | |
| SCVFZCLFOSHCOH-UHFFFAOYSA-M | |
| potassium;acetate | |
| 517044 | |
| 98.15 |
| 127-08-2 | |
| 1.57 | |
| MFCD00012458 | |
| 3595449 | |
| 14,7605 | |
| Soluble in water,alcohol and ammonia. Insoluble in acetone and ether. | |
| CC(=O)[O-].[K+] | |
| 98.142 | |
| CHEBI:32029 | |
| 99% |
Chemical Identifiers
| 127-08-2 | |
| 98.142 | |
| SCVFZCLFOSHCOH-UHFFFAOYSA-M | |
| 517044 | |
| potassium;acetate |
| C2H3KO2 | |
| MFCD00012458 | |
| potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 | |
| CHEBI:32029 | |
| CC(=O)[O-].[K+] |
Safety and Handling
H500
EINECSNumber : 204-822-2
RTECSNumber : AJ3325000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only