missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Acetate, ≥99%, MP Biomedicals
$53.71 - $207.56
Chemical Identifiers
| CAS | 127-08-2 |
|---|---|
| Molecular Formula | C2H3KO2 |
| Molecular Weight (g/mol) | 98.142 |
| InChI Key | SCVFZCLFOSHCOH-UHFFFAOYSA-M |
| Synonym | potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 |
| PubChem CID | 517044 |
| ChEBI | CHEBI:32029 |
| IUPAC Name | potassium;acetate |
| SMILES | CC(=O)[O-].[K+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN19484380
|
MP Biomedicals Inc
0219484380 |
100 g |
N/A
|
|
|||||
|
ICN19484391
|
MP Biomedicals Inc
0219484391 |
1 kg |
N/A
|
|
|||||
Chemical Identifiers
| 127-08-2 | |
| 98.142 | |
| potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 | |
| CHEBI:32029 | |
| CC(=O)[O-].[K+] |
| C2H3KO2 | |
| SCVFZCLFOSHCOH-UHFFFAOYSA-M | |
| 517044 | |
| potassium;acetate |
Specifications
| 127-08-2 | |
| ≥99% | |
| potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 | |
| CC(=O)[O-].[K+] | |
| 98.142 | |
| CHEBI:32029 | |
| Potassium Acetate |
| 100 g | |
| C2H3KO2 | |
| SCVFZCLFOSHCOH-UHFFFAOYSA-M | |
| potassium;acetate | |
| 517044 | |
| 98.14 |
Safety and Handling
Recommended Storage : Room Temperature