missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Acetate, ≥99%, MP Biomedicals
Specifications
Specifications
| Quantity | 100 g |
| Assay Percent Range | ≥99% |
| Formula Weight | 98.14 |
| Chemical Name or Material | Potassium Acetate |
Safety and Handling
| Recommended Storage | Room Temperature |