missing translation for 'onlineSavingsMsg'
Learn More

Potassium Acetate, ≥99.0%, ACS Reagent, Honeywell Fluka™

Catalog No. p-7120677
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60045442 100 g Poly Bottle
60045444 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60045442 Supplier Honeywell Chemicals Supplier No. 236497100G
Only null left

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 127-08-2
Molecular Formula C2H3KO2
Molecular Weight (g/mol) 98.142
MDL Number MFCD00012458
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
Synonym potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
PubChem CID 517044
ChEBI CHEBI:32029
IUPAC Name potassium;acetate
SMILES CC(=O)[O-].[K+]

Specifications

Melting Point 292°C
Physical Form Solid
Quantity 100 g
Linear Formula CH3COOK
UN Number NONH for all modes of transport
Beilstein 3595449
Solubility Information Soluble in water, 98.2g/L at 20°C
Formula Weight 98.14g/mol
Percent Purity ≥99.0%
Grade ACS Reagent
Packaging Poly Bottle
Density 1.57g/cu.cm at 25°C
Chemical Name or Material Potassium Acetate
Show More Show Less
EINECSNumber 204-822-2
RTECSNumber AJ3325000