missing translation for 'onlineSavingsMsg'
Learn More

Potassium acetate, 1M aq. soln., pH 7.5, RNAse free, Thermo Scientific Chemicals

$187.12 - $187.12

Chemical Identifiers

CAS 127-08-2
Molecular Formula C2H3KO2
Molecular Weight (g/mol) 98.142
MDL Number MFCD00012458
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
Synonym potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
PubChem CID 517044
ChEBI CHEBI:32029
IUPAC Name potassium;acetate
SMILES CC(=O)[O-].[K+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60832AK
View Documents
Thermo Scientific Chemicals
J60832AK
250 mL N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

127-08-2
98.142
SCVFZCLFOSHCOH-UHFFFAOYSA-M
517044
potassium;acetate
C2H3KO2
MFCD00012458
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
CHEBI:32029
CC(=O)[O-].[K+]

Specifications

Colorless
7.5
250 mL
CH3CO2K
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
CC(=O)[O-].[K+]
98.142
CHEBI:32029
1M aq. soln.
127-08-2
Liquid
C2H3KO2
MFCD00012458
SCVFZCLFOSHCOH-UHFFFAOYSA-M
potassium;acetate
517044
98.15
Potassium acetate, RNAse free
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only