missing translation for 'onlineSavingsMsg'
Learn More

Potassium acetate, 1M aq. soln., pH 7.5, RNAse free, Thermo Scientific Chemicals

Catalog No. AAJ60832AK
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60832AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ60832AK Supplier Thermo Scientific Chemicals Supplier No. J60832AK

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 127-08-2 | Molecular Formula: C2H3KO2 | Molecular Weight: 98.142 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 127-08-2
Molecular Formula C2H3KO2
Molecular Weight (g/mol) 98.142
MDL Number MFCD00012458
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
Synonym potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
PubChem CID 517044
ChEBI CHEBI:32029
IUPAC Name potassium;acetate
SMILES CC(=O)[O-].[K+]

Specifications

Color Colorless
pH 7.5
Physical Form Liquid
Quantity 250 mL
Linear Formula CH3CO2K
Formula Weight 98.15
Concentration or Composition (by Analyte or Components) 1M aq. soln.
Chemical Name or Material Potassium acetate, RNAse free
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only