missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium AA Standard, 1000 ppm K in 3% HCl, Ricca Chemical
Chemical Identifiers
| CAS | 7447-40-7 |
|---|---|
| Molecular Formula | ClK |
| Molecular Weight (g/mol) | 74.548 |
| InChI Key | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
| PubChem CID | 4873 |
| ChEBI | CHEBI:32588 |
| IUPAC Name | potassium;chloride |
| SMILES | [Cl-].[K+] |
| Catalog Number | Mfr. No. | Packaging | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Packaging | Price | Quantity | |||||
|
AK1KH-100
|
Ricca Chemical Company
AK1KH100 |
Natural LDPE Bottle |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AK1KH-500
|
Ricca Chemical Company
AK1KH500 |
Natural Poly Bottle |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 7447-40-7 | |
| 74.548 | |
| 4873 | |
| potassium;chloride |
| ClK | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| CHEBI:32588 | |
| [Cl-].[K+] |
Specifications
| Natural LDPE Bottle | |
| 95.0,3.05,0.16 | |
| ClK | |
| [Cl-].[K+] | |
| 74.548 | |
| CHEBI:32588 | |
| Potassium AA Standard |
| 7447-40-7,7647-01-0,7447-40-7 | |
| 99.0,3.18,0.17 | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| potassium;chloride | |
| 4873 | |
| 1mL = 1mg K (1, 000ppm K)KCl in 3% HCl |
Safety and Handling
ShelfLife : 24 months