Learn More
Potassium 4-methylphenyltrifluoroborate, 98%
CAS: 216434-82-1 | C7H7BF3K | 198.037 g/mol
Supplier: Thermo Scientific Chemicals L1760406
Description
In Suzuki Cross-Coupling organotrifluoroborates is used as potent boronic acid surrogates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIn Suzuki Cross-Coupling organotrifluoroborates is used as potent boronic acid surrogates.
Solubility
Soluble in water.
Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
Specifications
| Potassium 4-methylphenyltrifluoroborate | |
| >300°C | |
| MFCD01318171 | |
| 8290098 | |
| Soluble in water. | |
| [B-](C1=CC=C(C=C1)C)(F)(F)F.[K+] | |
| 198.037 | |
| 198.04 |
| 216434-82-1 | |
| C7H7BF3K | |
| 5 g | |
| potassium p-tolyltrifluoroborate, potassium trifluoro p-tolyl borate, potassium 4-methylphenyl trifluoroborate, potassium 4-methylphenyltrifluoroborate, potassium trifluoro 4-methylphenyl boranuide, 4-methylphenyltrifluoroboric acid potassium salt, pubchem11452, trifluoro 4-methylphenyl potassioboron v, potassium trifluoro p-tolyl boranuide, potassium trifluoro-4-methylphenyl boranuide | |
| KRWDYXJWQBTBAH-UHFFFAOYSA-N | |
| potassium;trifluoro-(4-methylphenyl)boranuide | |
| 23682258 | |
| 98% |
Chemical Identifiers
| 216434-82-1 | |
| 198.037 | |
| KRWDYXJWQBTBAH-UHFFFAOYSA-N | |
| 23682258 | |
| [B-](C1=CC=C(C=C1)C)(F)(F)F.[K+] |
| C7H7BF3K | |
| MFCD01318171 | |
| potassium p-tolyltrifluoroborate, potassium trifluoro p-tolyl borate, potassium 4-methylphenyl trifluoroborate, potassium 4-methylphenyltrifluoroborate, potassium trifluoro 4-methylphenyl boranuide, 4-methylphenyltrifluoroboric acid potassium salt, pubchem11452, trifluoro 4-methylphenyl potassioboron v, potassium trifluoro p-tolyl boranuide, potassium trifluoro-4-methylphenyl boranuide | |
| potassium;trifluoro-(4-methylphenyl)boranuide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only