missing translation for 'onlineSavingsMsg'
Learn More

Potassium 4-methylphenyltrifluoroborate, 98%

Catalog No. p-7047101
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1760403 1 g
AAL1760406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1760403 Supplier Thermo Scientific Chemicals Supplier No. L1760403
Only null left

CAS: 216434-82-1 | C7H7BF3K | 198.037 g/mol

In Suzuki Cross-Coupling organotrifluoroborates is used as potent boronic acid surrogates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In Suzuki Cross-Coupling organotrifluoroborates is used as potent boronic acid surrogates.

Solubility
Soluble in water.

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 216434-82-1
Molecular Formula C7H7BF3K
Molecular Weight (g/mol) 198.037
MDL Number MFCD01318171
InChI Key KRWDYXJWQBTBAH-UHFFFAOYSA-N
Synonym potassium p-tolyltrifluoroborate, potassium trifluoro p-tolyl borate, potassium 4-methylphenyl trifluoroborate, potassium 4-methylphenyltrifluoroborate, potassium trifluoro 4-methylphenyl boranuide, 4-methylphenyltrifluoroboric acid potassium salt, pubchem11452, trifluoro 4-methylphenyl potassioboron v, potassium trifluoro p-tolyl boranuide, potassium trifluoro-4-methylphenyl boranuide
PubChem CID 23682258
IUPAC Name potassium;trifluoro-(4-methylphenyl)boranuide
SMILES [B-](C1=CC=C(C=C1)C)(F)(F)F.[K+]

Specifications

Melting Point >300°C
Quantity 1 g
Beilstein 8290098
Solubility Information Soluble in water.
Formula Weight 198.04
Percent Purity 98%
Chemical Name or Material Potassium 4-methylphenyltrifluoroborate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only