missing translation for 'onlineSavingsMsg'
Learn More

Potassium 4-fluorophenyltrifluoroborate, 95%

Catalog No. AAL1765506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1765506 5 g
AAL1765503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1765506 Supplier Thermo Scientific Chemicals Supplier No. L1765506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 192863-35-7 | C6H4BF4K | 202 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 192863-35-7
Molecular Formula C6H4BF4K
Molecular Weight (g/mol) 202
MDL Number MFCD01318170
InChI Key PXKNUQDMYBUYSZ-UHFFFAOYSA-N
Synonym potassium 4-fluorophenyltrifluoroborate, potassium trifluoro 4-fluorophenyl borate, potassium trifluoro 4-fluorophenyl boranuide, potassium 4-fluorophenyl trifluoroborate, 4-fluorophenyl-trifluoroborate potassium salt, 4-fluorophenyltrifluoroboric acid potassium salt, 4-fluorobenzenetrifluoroboric acid potassium salt, pubchem11422, potassiotrifluoro 4-fluorophenyl boron v, potassium trifluoro-4-fluorophenyl boranuide
PubChem CID 23677644
IUPAC Name potassium;trifluoro-(4-fluorophenyl)boranuide
SMILES [B-](C1=CC=C(C=C1)F)(F)(F)F.[K+]

Specifications

Melting Point 290°C to 292°C
Quantity 5 g
Beilstein 7783544
Formula Weight 202
Percent Purity 95%
Chemical Name or Material Potassium 4-fluorophenyltrifluoroborate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only