missing translation for 'onlineSavingsMsg'
Learn More

Potassium 3-bromophenyltrifluoroborate, 97%

Catalog No. AAL1796603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1796603 1 g
AAL1796606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1796603 Supplier Thermo Scientific Chemicals Supplier No. L1796603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 374564-34-8 | C6H4BBrF3K | 262.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 374564-34-8
Molecular Formula C6H4BBrF3K
Molecular Weight (g/mol) 262.91
MDL Number MFCD02093977
InChI Key NIDYRVJZTADCGO-UHFFFAOYSA-N
Synonym potassium 3-bromophenyl trifluoroborate, potassium 3-bromophenyltrifluoroborate, potassium 3-bromophenyl trifluoroboranuide, amtb081, potassium 3-bromophenyl-trifluoroboranuide, potassium ion 3-bromophenyl trifluoroboranuide, potassium 3-bromophenyl trifluoro borate 1-, potassium 3-bromophenyl-tris fluoranyl boranuide
PubChem CID 23677642
IUPAC Name potassium;(3-bromophenyl)-trifluoroboranuide
SMILES [K+].F[B-](F)(F)C1=CC(Br)=CC=C1

Specifications

Melting Point 178°C to 182°C
Quantity 1 g
Formula Weight 262.91
Percent Purity 97%
Chemical Name or Material Potassium 3-bromophenyltrifluoroborate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only