missing translation for 'onlineSavingsMsg'
Learn More

Potassium 2-phenylethyltrifluoroborate, 98%

Catalog No. p-7028549
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2593003 1 g
AAH2593006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2593003 Supplier Thermo Scientific Chemicals Supplier No. H2593003

CAS: 329976-74-1 | C8H9BF3K | 212.064 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 329976-74-1
Molecular Formula C8H9BF3K
Molecular Weight (g/mol) 212.064
MDL Number MFCD09039257
InChI Key SOZVPVAYKGJPJS-UHFFFAOYSA-N
Synonym potassium phenethyltrifluoroborate, potassium trifluoro phenethyl borate, potassium trifluoro 2-phenylethyl boranuide, pubchem11290, potassiumphenethyltrifluoroborate, trifluoropotassiophenethylboron v, potassium 2-phenethyltrifluoroborate, potassium 2-phenylethyltrifluoroborate, potassium trifluoro 1-phenylethyl borate
PubChem CID 23697312
IUPAC Name potassium;trifluoro(2-phenylethyl)boranuide
SMILES [B-](CCC1=CC=CC=C1)(F)(F)F.[K+]

Specifications

Melting Point 173°C to 175°C
Quantity 1 g
Formula Weight 212.06
Percent Purity 98%
Chemical Name or Material Potassium 2-phenylethyltrifluoroborate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only