missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium 2-methylthiophenyltrifluoroborate, 96%, Thermo Scientific™
$215.37 - $215.37
Chemical Identifiers
| CAS | 850623-77-7 |
|---|---|
| Molecular Formula | C7H7BF3KS |
| Molecular Weight (g/mol) | 230.097 |
| MDL Number | MFCD04115773 |
| InChI Key | ACOYJOWMHXJLJP-UHFFFAOYSA-N |
| Synonym | potassium trifluoro 2-methylthio phenyl borate, potassium 2-methylthiophenyl trifluoroborate, potassium trifluoro 2-methylsulfanyl phenyl boranuide, potassium 2-methylsulfanylphenyl trifluoroborate, potassium 2-methylthiophenyltrifluoroborate, potassium trifluoro-2-methylthio phenyl boranuide, potassium trifluoro 2-methylsulfanyl phenyl borate 1-, potassium ion trifluoro 2-methylsulfanyl phenyl boranuide, potassium tris fluoranyl-2-methylsulfanylphenyl boranuide |
| PubChem CID | 44717213 |
| IUPAC Name | potassium;trifluoro-(2-methylsulfanylphenyl)boranuide |
| SMILES | [B-](C1=CC=CC=C1SC)(F)(F)F.[K+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5258703
|
Thermo Scientific Chemicals
H5258703 |
1 g |
Each for $215.37
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 850623-77-7 | |
| 230.097 | |
| ACOYJOWMHXJLJP-UHFFFAOYSA-N | |
| 44717213 | |
| [B-](C1=CC=CC=C1SC)(F)(F)F.[K+] |
| C7H7BF3KS | |
| MFCD04115773 | |
| potassium trifluoro 2-methylthio phenyl borate, potassium 2-methylthiophenyl trifluoroborate, potassium trifluoro 2-methylsulfanyl phenyl boranuide, potassium 2-methylsulfanylphenyl trifluoroborate, potassium 2-methylthiophenyltrifluoroborate, potassium trifluoro-2-methylthio phenyl boranuide, potassium trifluoro 2-methylsulfanyl phenyl borate 1-, potassium ion trifluoro 2-methylsulfanyl phenyl boranuide, potassium tris fluoranyl-2-methylsulfanylphenyl boranuide | |
| potassium;trifluoro-(2-methylsulfanylphenyl)boranuide |
Specifications
| 850623-77-7 | |
| C7H7BF3KS | |
| 1 g | |
| ACOYJOWMHXJLJP-UHFFFAOYSA-N | |
| potassium;trifluoro-(2-methylsulfanylphenyl)boranuide | |
| 44717213 | |
| 96% |
| 274°C to 276°C | |
| MFCD04115773 | |
| potassium trifluoro 2-methylthio phenyl borate, potassium 2-methylthiophenyl trifluoroborate, potassium trifluoro 2-methylsulfanyl phenyl boranuide, potassium 2-methylsulfanylphenyl trifluoroborate, potassium 2-methylthiophenyltrifluoroborate, potassium trifluoro-2-methylthio phenyl boranuide, potassium trifluoro 2-methylsulfanyl phenyl borate 1-, potassium ion trifluoro 2-methylsulfanyl phenyl boranuide, potassium tris fluoranyl-2-methylsulfanylphenyl boranuide | |
| [B-](C1=CC=CC=C1SC)(F)(F)F.[K+] | |
| 230.097 | |
| 230.1 | |
| Potassium 2-methylthiophenyltrifluoroborate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures