missing translation for 'onlineSavingsMsg'
Learn More

Potassium 1-Naphthaleneacetate 99.0+%, TCI America™

Catalog No. N000625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N000625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N000625G Supplier TCI America Supplier No. N000625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15165-79-4
Molecular Formula C12H9KO2
Molecular Weight (g/mol) 224.30
MDL Number MFCD00067168
InChI Key HPQBUYIHTJNBOM-UHFFFAOYSA-M
Synonym 1-Naphthaleneacetic Acid Potassium Salt, 1-Naphthylacetic Acid Potassium Salt, Potassium 1-Naphthylacetate, K-NAA
PubChem CID 23700091
IUPAC Name potassium 2-(naphthalen-1-yl)acetate
SMILES [K+].[O-]C(=O)CC1=C2C=CC=CC2=CC=C1

Specifications

Color White
Quantity 25 g
Formula Weight 224.30
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium 1-Naphthaleneacetate
RTECSNumber QJ1270000
TSCA No