accessibility menu, dialog, popup

UserName

CAS RN 15165-79-4

IUPAC Name: potassium 2-(naphthalen-1-yl)acetate
Synonyms: potassium 2-naphthalen-1-ylacetate
Molecular Weight (g/mol): 224.3
Molecular Formula: C12H9KO2
InChi Key: HPQBUYIHTJNBOM-UHFFFAOYSA-M
SMILES: [K+].[O-]C(=O)CC1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results