missing translation for 'onlineSavingsMsg'
Learn More

Potassium Sulfate, (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. P3043
Change view
Click to view available options
Quantity:
3 kg
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
P3043 3 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P3043 Supplier Thermo Fisher Scientific Supplier No. P304 3
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7778-80-5
Molecular Formula K2O4S
Molecular Weight (g/mol) 174.25
MDL Number MFCD00011388
InChI Key OTYBMLCTZGSZBG-UHFFFAOYSA-L
Synonym potassium sulfate
PubChem CID 24507
ChEBI CHEBI:32036
IUPAC Name dipotassium sulfate
SMILES [K+].[K+].[O-]S([O-])(=O)=O

Specifications

Boiling Point 1689°C
Melting Point 1,067°C
Color White
pH 6 to 8
Physical Form Solid
Quantity 3 kg
Formula Weight 174.26
Percent Purity ≥99%
Grade Certified ACS
Identification Pass Test
Packaging Poly Bottle
Assay ≥99 %
Chemical Name or Material Potassium Sulfate
Show More Show Less