missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ POROS™ MabCapture™ A RoboColumn™ Pre-packed columns
GREENER_CHOICE

Catalog No. A30725
Change view
Click to view available options
Quantity:
200 μL Column
600 μL Column
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30725 200 μL Column
A30726 600 μL Column
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A30725 Supplier Thermo Scientific™ Supplier No. A30725
Only null left

Thermo Scientific™ RoboColumn™ chromatography products are small columns provided in 8-column strips.

RoboColumn™ chromatography products are small columns provided in 8-column strips. They are useful for fully automated and parallel chromatographic separations using robotic liquid handling platforms. These particular RoboColumns contain POROS MabCapture A affinity resin, which is suitable for monoclonal antibody large-scale downstream bioprocessing.

POROS MabCapture A is a 50-micron affinity resin that combines improvements made in bead and chemistry technology. These advances provide a monoclonal antibody purification resin that not only demonstrates the highest dynamic binding performance at low flow rates, but also one that can maintain superior binding capacity at high linear velocities where other chromatography resins products cannot operate.

Features of the resin:

  • Obtain the highest dynamic binding capacity for monoclonal antibodies vs. conventional agarose or silica based Protein A medias
  • Maintain high dynamic binding capacity at flow rates over 700 cm/hr
  • Design processes with greater flexibility (shorter bed heights, faster flow rates or both) due to the improvements made to the antibody binding efficiency
  • Clean and sanitize with standard sodium hydroxide agents utilized for other chromatography products

RoboColumn™ is a registered trademark of ATOLL GmbH.

Specifications

Product Type Column
Product Line MABCAPTURE, POROS
Column Type Affinity
Format Chromatography Cartridge
Stationary Phase Protein A
Purification Target Protein
Content And Storage Store at 2°C to 8°C.
Quantity 200 μL Column
Shipping Condition Wet Ice