missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Porcine TGF alpha ELISA Kit
ELISA
Supplier: Invitrogen™ ES21RB
Description
Porcine TGF alpha quantitates porcine TGF alpha in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant porcine TGF alpha.
TGF-alpha is an EGF-related polypeptide growth factor that signals through the EGF receptor, and stimulates the proliferation of a wide range of epidermal and epithelial cells. It is produced by monocytes, keratinocytes, and various tumor cells. TGF-alpha induces anchorage-independence transformation in cultured cells. Human, murine and rat TGF-alpha are cross-species reactive. TGFalpha (aa 50) is a growth factor with 33% homology to EGF, binds to EGFR, activates tyrosine phosphorylation of the receptor, and stimulates cell proliferation. It plays a role in tumor initiation by inducing the reversible transformed phenotype.Chemical Identifiers
| EGF-like TGF; ETGF; LOC100009482; protransforming growth factor alpha; RATTGFAA; TFGA; TGF a; TGF type 1; TGF a; Tgfa; TGFAA; TGFalpha; TGF-alpha; TGFa; Transforming growth factor; Transforming growth factor alpha; transforming growth factor, alpha; transforming growth factor-alpha; wa1; wa-1; waved 1 | |
| 0.6 pg/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| TGFA | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| TGF alpha | |
| 4 hrs 45 mins |
| 0.6 to 120 pg/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 397484 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |
Specifications
| EGF-like TGF; ETGF; LOC100009482; protransforming growth factor alpha; RATTGFAA; TFGA; TGF a; TGF type 1; TGF a; Tgfa; TGFAA; TGFalpha; TGF-alpha; TGFa; Transforming growth factor; Transforming growth factor alpha; transforming growth factor, alpha; transforming growth factor-alpha; wa1; wa-1; waved 1 | |
| 0.6 pg/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| TGFA | |
| <10 | |
| HRP | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |
| 0.6 to 120 pg/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 397484 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| 96 Tests | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| TGF alpha | |
| 4 hrs 45 mins |