missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Porcine OPG ELISA Kit

Catalog No. ES19RBX5
Encompass
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES19RBX5 5 x 96 Tests
ES19RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ES19RBX5 Supplier Invitrogen™ Supplier No. ES19RBX5
Only null left

ELISA

Porcine OPG quantitates porcine OPG in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant porcine OPG.

The protein encoded by this gene is a member of the TNF-receptor superfamily. This protein is an osteoblast-secreted decoy receptor that functions as a negative regulator of bone resorption. This protein specifically binds to its ligand, osteoprotegerin ligand, both of which are key extracellular regulators of osteoclast development. Studies of the mouse counterpart also suggest that this protein and its ligand play a role in lymph-node organogenesis and vascular calcification. Alternatively spliced transcript variants of this gene have been reported, but their full length nature has not been determined.

Specifications

Accession Number LOW QUALITY PROTEIN: tumor necrosis factor receptor superfamily member 11 B; MGC29565; OCIF; Opg; Osteoclastogenesis inhibitory factor; Osteoprotegerin; PDB5; soluble OPG; soluble Osteoprotegerin; sOPG; TNF receptor superfamily member 11 b; Tnfrsf11b; TR1; Tumor necrosis factor receptor superfamily member 11 B; tumor necrosis factor receptor superfamily, member 11 b; tumor necrosis factor receptor superfamily, member 11 b (osteoprotegerin)
Assay Range 0.1 to 30 ng/mL
Assay Sensitivity 0.12 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 100049688
Gene Symbol TNFRSF11B
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target TNFRSF11B
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less