missing translation for 'onlineSavingsMsg'
Learn More

Porcine IL-6 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Catalog No. IC6861S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC6861S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IC6861S100 Supplier Bio-Techne Supplier No. IC6861S100UG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

IL-6 Monoclonal specifically detects IL-6 in Pig samples. It is validated for Flow Cytometry (Intracellular Staining).

Specifications

Antigen IL-6
Applications Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 90613
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 750
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P26893
Gene Alias B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A
Gene Symbols IL6
Host Species Mouse
Immunogen E. coli-derived recombinant porcine IL-6 Pro29-Met212 Accession # P26893
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3569
Test Specificity Detects porcine IL-6 in direct ELISAs and Western blots. In direct ELISAs and Western blots, partial cross-reactivity with recombinant human, mouse, canine, equine, and feline IL-6 is observed, and no cross-reactivity with recombinant human (rh) CLC, rhIL-11, rhCNTF, rhCT-1, rhLIF, or rhOSM is observed.
Target Species Pig
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Product Type Antibody
Isotype IgG2b
Product Line R&D Systems
Show More Show Less

For Research Use Only (RUO)