missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ POP-7™ Polymer for Protein Quality Analysis

Catalog No. p-7138023
Encompass
Change view
Click to view available options
Quantity:
384 Samples
960 Samples
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30936 384 Samples
A31122 960 Samples
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A30936 Supplier Applied Biosystems™ Supplier No. A30936
Only null left

Includes

Radio frequency identification (RFID) label.

This is a pouch of POP-7 polymer for protein quality analysis on 3500/3500xL genetic analyzers. The POP-7 separation matrix is validated for protein quality analysis and is offered in easy-to-use pouch packages and 960 sample (maximum) or 384 sample (maximum) size formats.

  • Easy to use: Pre-formulated and in a single-use pouch, which not only saves time but ensures reliability and consistency
  • Robust formulation: Formulated for biomolecular analysis applications
  • Replenishable: Allows capillaries to be used multiple times
  • Includes radio frequency identification (RFID) label: Easily track important information such as part/lot number, samples remaining, and expiry dates (displayed on software dashboard)

Specifications

Content And Storage Store at 2–8°C.
Description POP-7™ Polymer For 3500xL Genetic Analyzer
For Use With (Equipment) 3500xL Genetic Analyzer for Protein Quality Analysis
For Use With (Application) Glycan Analysis, Protein Quality Analysis
No. of Samples 384
Product Line POP-7
Product Type Polymer
Quantity 384 Samples
Shipping Condition Wet Ice

For research use only