missing translation for 'onlineSavingsMsg'
Learn More

Pop-4 Bottle (960) with RFID For 3500 Genetic Analyzer

Catalog No. 4482242
Encompass
Change view
Click to view available options
Quantity:
960 Samples
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4482242 960 Samples
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4482242 Supplier Thermo Fisher Scientific Supplier No. 4482242

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Convenient Pop-4 Bottle with RFID for use with the 3500 Genetic Analyzer

Pop-4 Bottle (960) with RFID For 3500 Genetic Analyzer

Order Info

Each

Certifications

ISO 9001 and ISO 13485

Specifications

Description Pop-4 Bottle (960), For 3500 Genetic Analyzer, With RFID
For Use With (Equipment) 3500 Genetic Analyzer
Includes RFID
No. of Samples 960
Product Type POP-4™ Polymer
Quantity 960 Samples
Sample Volume 960 Sample
Size 7mL

For research use only