missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ponceau Xylidine, ≥96.0% (HPLC), MilliporeSigma™ Supelco™
Description
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications
Specifications
| CAS | 3761-53-3 |
| Physical Form | Neat |
| Linear Formula | C18H14N2 Na2O7S2 |
| MDL Number | MFCD00003897 |
| Synonym | Acid Red 26; Ponceau 2 R; Xylidine ponceau; Xylidine ponceau 2 R |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 480.42 |
| Percent Purity | ≥96.0% (HPLC) |
| Grade | Analytical Standard |