missing translation for 'onlineSavingsMsg'
Learn More

Ponceau S, 0.1% v/v soln. in 5% acetic acid

CAS: 6226-79-5 | C22H12N4Na4O13S4 | 760.552 g/mol

$187.12 - $315.99

Chemical Identifiers

CAS 6226-79-5
Molecular Formula C22H12N4Na4O13S4
Molecular Weight (g/mol) 760.552
MDL Number MFCD00003892
InChI Key VSXKEUCERCWGKF-STNZDNLRSA-J
Synonym ponceau s, acid red 112, ponceau s extra, ponceau red s, unii-t1c2ugs05f, c.i. acid red 112, t1c2ugs05f, 3-hydroxy-4-2-sulfo-4-4-sulfophenylazo phenylazo-2,7-naphthalenedisulfonic acid sodium salt, c.i. acid red 112, tetrasodium salt, ponceau-s
PubChem CID 11320219
IUPAC Name tetrasodium;(4Z)-3-oxo-4-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalene-2,7-disulfonate
SMILES C1=CC(=CC=C1N=NC2=CC(=C(C=C2)NN=C3C4=C(C=C(C=C4)S(=O)(=O)[O-])C=C(C3=O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ63139AP
View Documents
Thermo Scientific Chemicals
J63139AP
500 mL
Each for $187.12
Only null left
 
AAJ63139K2
View Documents
Thermo Scientific Chemicals
J63139K2
1 L
Each for $315.99
Only null left
 
Description

Description

Ponceau S is used to prepare a stain for rapid reversible detection of protein bands on nitrocellulose or polyvinylidene fluoride membranes, as well as on cellulose acetate membranes. And it is also used as a rapid protein band stain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6226-79-5
760.552
VSXKEUCERCWGKF-STNZDNLRSA-J
11320219
C1=CC(=CC=C1N=NC2=CC(=C(C=C2)NN=C3C4=C(C=C(C=C4)S(=O)(=O)[O-])C=C(C3=O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+]
C22H12N4Na4O13S4
MFCD00003892
ponceau s, acid red 112, ponceau s extra, ponceau red s, unii-t1c2ugs05f, c.i. acid red 112, t1c2ugs05f, 3-hydroxy-4-2-sulfo-4-4-sulfophenylazo phenylazo-2,7-naphthalenedisulfonic acid sodium salt, c.i. acid red 112, tetrasodium salt, ponceau-s
tetrasodium;(4Z)-3-oxo-4-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalene-2,7-disulfonate

Specifications

Red
C22H12N4Na4O13S4
0.2% v/v soln. in 5% acetic acid
Soluble in water (1mg/ml). Insoluble in ether.
C1=CC(=CC=C1N=NC2=CC(=C(C=C2)NN=C3C4=C(C=C(C=C4)S(=O)(=O)[O-])C=C(C3=O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+]
760.552
760.56
Liquid
6226-79-5
MFCD00003892
ponceau s, acid red 112, ponceau s extra, ponceau red s, unii-t1c2ugs05f, c.i. acid red 112, t1c2ugs05f, 3-hydroxy-4-2-sulfo-4-4-sulfophenylazo phenylazo-2,7-naphthalenedisulfonic acid sodium salt, c.i. acid red 112, tetrasodium salt, ponceau-s
VSXKEUCERCWGKF-STNZDNLRSA-J
tetrasodium;(4Z)-3-oxo-4-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalene-2,7-disulfonate
11320219
500 mL
Ponceau S
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only