missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ponceau S, 0.1% v/v soln. in 5% acetic acid
Description
Ponceau S is used to prepare a stain for rapid reversible detection of protein bands on nitrocellulose or polyvinylidene fluoride membranes, as well as on cellulose acetate membranes. And it is also used as a rapid protein band stain.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Red |
| Concentration | 0.2% v/v soln. in 5% acetic acid |
| Solubility Information | Soluble in water (1mg/ml). Insoluble in ether. |
| Formula Weight | 760.56 |
| Quantity | 500 mL |
| Physical Form | Liquid |
| Chemical Name or Material | Ponceau S |
Safety and Handling
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only