missing translation for 'onlineSavingsMsg'
Learn More

Ponceau 6R, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013801
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013801 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013801 Supplier Merck Emd Millipore Supplier No. 9636525MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ponceau 6R is a red azo dye which is widely used as a food colorant and also used in histology staining. It is soluble in water and slightly soluble in ethanol.

Specifications

CAS 2766-77-0
Physical Form Neat
Linear Formula C20H12N2 Na2O7S2
MDL Number MFCD00013279
Melting Point >300°C
Synonym Crystal Ponceau 6 R; Acid Red 44; Brilliant Crystal Scarlet; Crystal Scarlet; Ponceau 6 R
Type Analytical Standard
Quantity 25 mg
Formula Weight 502.43
Percent Purity ≥98% (HPLC)
Grade Analytical Standard
Show More Show Less