missing translation for 'onlineSavingsMsg'
Learn More

Ponceau 4R, Analytical Standard, ≥99.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013767
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013767 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013767 Supplier Merck Emd Millipore Supplier No. 1813725MG
Only null left

Ponceau 4R is a synthetic red azo dye and is used as a food colorant. It is used in range of alcoholic and non-alcoholic beverages, food stuffs, preserved fruits etc. It exists as trisodium-2-hydroxy-1-(4-sulfonato-1-naphthylazo)-6,8-naphthalenedisulfonate.

Specifications

Packaging Bottomless Glass Bottle with Fused Cone
CAS 2611-82-7
Physical Form Neat
Linear Formula C20H11N2Na3O10S3
MDL Number MFCD00004084
Synonym New Coccine; Acid Red 18; Ponceau 4 R
Quantity 25 mg
Formula Weight 604.47
Percent Purity ≥99.0% (HPLC)
Grade Analytical Standard