missing translation for 'onlineSavingsMsg'
Learn More

Pomalidomide 98.0+%, TCI America™

Supplier:  TCI America P2074100MG

 View more versions of this product

Catalog No. P2074100MG


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Pomalidomide
313°C
C13H11N3O4
100 mg
UVSMNLNDYGZFPF-UHFFFAOYNA-N
4-amino-2-(2,6-dioxopiperidin-3-yl)-2,3-dihydro-1H-isoindole-1,3-dione
134780
273.25
Crystalline Powder
19171-19-8
Yellow
MFCD12756407
3-Amino-N-(2,6-dioxo-3-piperidyl)phthalimide
NC1=CC=CC2=C1C(=O)N(C1CCC(=O)NC1=O)C2=O
273.25
CHEBI:72690
≥98.0% (HPLC)

Chemical Identifiers

19171-19-8
273.25
UVSMNLNDYGZFPF-UHFFFAOYNA-N
134780
4-amino-2-(2,6-dioxopiperidin-3-yl)-2,3-dihydro-1H-isoindole-1,3-dione
C13H11N3O4
MFCD12756407
3-Amino-N-(2,6-dioxo-3-piperidyl)phthalimide
CHEBI:72690
NC1=CC=CC2=C1C(=O)N(C1CCC(=O)NC1=O)C2=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : NR3397905

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions