missing translation for 'onlineSavingsMsg'
Learn More

Pomalidomide 98.0+%, TCI America™

Catalog No. P207425MG
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P207425MG 25 mg
P2074100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P207425MG Supplier TCI America Supplier No. P207425MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19171-19-8
Molecular Formula C13H11N3O4
Molecular Weight (g/mol) 273.25
MDL Number MFCD12756407
InChI Key UVSMNLNDYGZFPF-UHFFFAOYNA-N
Synonym 3-Amino-N-(2,6-dioxo-3-piperidyl)phthalimide
PubChem CID 134780
ChEBI CHEBI:72690
IUPAC Name 4-amino-2-(2,6-dioxopiperidin-3-yl)-2,3-dihydro-1H-isoindole-1,3-dione
SMILES NC1=CC=CC2=C1C(=O)N(C1CCC(=O)NC1=O)C2=O

Specifications

Melting Point 313°C
Color Yellow
Quantity 25 mg
Formula Weight 273.25
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Pomalidomide
RTECSNumber NR3397905
TSCA No
Recommended Storage Refrigerator