missing translation for 'onlineSavingsMsg'
Learn More

Polyvinyl Alcohol, White to off-white powder, MP Biomedicals

Catalog No. ICN15193883
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15193883 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15193883 Supplier MP Biomedicals Inc Supplier No. 0215193883
Only null left

Chemical Identifiers

CAS 9002-89-5
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 44.05
MDL Number MFCD00081922
InChI Key IMROMDMJAWUWLK-UHFFFAOYSA-N
Synonym vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval
PubChem CID 11199
IUPAC Name ethenol
SMILES OC(-*)C-*

Specifications

Color White
Density 1.25-1.50g/mL
Assay Percent Range 4%
Quantity 250 g
Formula Weight 22000
Physical Form Powder
Chemical Name or Material Polyvinyl Alcohol, Aqueous Solution