missing translation for 'onlineSavingsMsg'
Learn More

Polyvinyl Alcohol, USP, Spectrum™ Chemical

Catalog No. 18605413
Change view
Click to view available options
Quantity:
1 kg
12 kg
25 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605413 25 kg Fiber Drum
18605409 1 kg Poly Bottle
18605410 12 kg Poly Pail
18605411 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18605413 Supplier Spectrum Chemical Mfg Cor Supplier No. P118025KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

P1180, 9002-89-5, (C2H4O)n

Spectrum™ Chemical Polyvinyl Alcohol, USP is used to relieve dry, irritated eyes. All Spectrum Chemical USP are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9002-89-5
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 44.05
MDL Number MFCD00081922
InChI Key IMROMDMJAWUWLK-UHFFFAOYSA-N
IUPAC Name ethenol
SMILES OC(-*)C-*

Specifications

pH 5.0 to 8.0
Acid Value 3
CAS Max % 1
Quantity 25 kg
Grade USP
Ignition Residue 0.01
Loss on Drying 0.05
Packaging Fiber Drum