missing translation for 'onlineSavingsMsg'
Learn More

Polyvinyl Alcohol, Hydrolyzed, Reagent, 99-100%, Spectrum™ Chemical

Catalog No. p-7158034
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
25 kg
Packaging:
Fiber Drum
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605402 1 kg Poly Pail
18605403 2.5 kg Poly Pail
18605404 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18605402 Supplier Spectrum Chemical Mfg Cor Supplier No. PO1411KG
Only null left

PO141, 9002-89-5, (C2H4O)n

Spectrum™ Chemical Polyvinyl Alcohol, 99-100 Percent, Hydrolyzed, Reagent , also known as PVA or PVAI, is a synthetic polymer used in textiles, papermaking, and several types of coatings. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 9002-89-5
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 44.05
MDL Number MFCD00081922
InChI Key IMROMDMJAWUWLK-UHFFFAOYSA-N
IUPAC Name ethenol
SMILES OC(-*)C-*

Specifications

CAS Max % 1
Quantity 1 kg
Grade Reagent
Packaging Poly Pail
Viscosity 24-32 cps