Learn More
Polyvinyl alcohol, 98-99% hydrolyzed, medium molecular weight
CAS: 9002-89-5 | (C2H4O)n | 44.05 g/mol
$60.01 - $476.63
Chemical Identifiers
| CAS | 9002-89-5 |
|---|---|
| Molecular Formula | (C2H4O)n |
| Molecular Weight (g/mol) | 44.05 |
| MDL Number | MFCD00081922 |
| InChI Key | IMROMDMJAWUWLK-UHFFFAOYSA-N |
| Synonym | vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval |
| PubChem CID | 11199 |
| IUPAC Name | ethenol |
| SMILES | OC(-*)C-* |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA4124214
|
Thermo Scientific Chemicals
04124214 |
25 g |
Each for $60.01
|
|
|||||
|
AA4124222
|
Thermo Scientific Chemicals
04124222 |
100 g |
Each for $122.85
|
|
|||||
|
AA4124236
|
Thermo Scientific Chemicals
04124236 |
500 g |
Each for $476.63
|
|
|||||
Description
Polyvinyl alcohol is used in papermaking, textiles, and in a variety of coatings. It is used as an emulsion polymerization aid to make polyvinyl acetate, as a protective colloid, as a film used in the water transfer printing process, as a surfactant for the formation of polymer encapsulated nanobeads, as an embolization agent in medical procedures and as a fixative for specimen collection. It is an important raw material for polyvinyl nitrate (PVN). It is used in protective chemical-resistant gloves, in eye drops (such as artificial tears to treat dry eyes) and in hard contact lens solution as a lubricant and in 3D printing as a support structure that can then be dissolved away.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 9002-89-5 | |
| 44.05 | |
| IMROMDMJAWUWLK-UHFFFAOYSA-N | |
| 11199 | |
| OC(-*)C-* |
| (C2H4O)n | |
| MFCD00081922 | |
| vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval | |
| ethenol |
Specifications
| 9002-89-5 | |
| MFCD00081922 | |
| Soluble in water. Slightly soluble in ethanol. Insoluble in other organic solvents. | |
| IMROMDMJAWUWLK-UHFFFAOYSA-N | |
| ethenol | |
| 11199 | |
| Odorless |
| (C2H4O)n | |
| vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval | |
| 25 g | |
| OC(-*)C-* | |
| 44.05 | |
| 98 to 99% | |
| Polyvinyl alcohol, Medium molecular weight, Hydrolyzed |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only