missing translation for 'onlineSavingsMsg'
Learn More

Polyvinyl alcohol, 98-99% hydrolyzed, medium molecular weight

Catalog No. AA4124214
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4124214 25 g
AA4124222 100 g
AA4124236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4124214 Supplier Thermo Scientific Chemicals Supplier No. 04124214

CAS: 9002-89-5 | (C2H4O)n | 44.05 g/mol

Polyvinyl alcohol is used in papermaking, textiles, and in a variety of coatings. It is used as an emulsion polymerization aid to make polyvinyl acetate, as a protective colloid, as a film used in the water transfer printing process, as a surfactant for the formation of polymer encapsulated nanobeads, as an embolization agent in medical procedures and as a fixative for specimen collection. It is an important raw material for polyvinyl nitrate (PVN). It is used in protective chemical-resistant gloves, in eye drops (such as artificial tears to treat dry eyes) and in hard contact lens solution as a lubricant and in 3D printing as a support structure that can then be dissolved away.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9002-89-5
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 44.05
MDL Number MFCD00081922
InChI Key IMROMDMJAWUWLK-UHFFFAOYSA-N
Synonym vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval
PubChem CID 11199
IUPAC Name ethenol
SMILES OC(-*)C-*

Specifications

Solubility Information Soluble in water. Slightly soluble in ethanol. Insoluble in other organic solvents.
Quantity 25 g
Percent Purity 98 to 99%
Odor Odorless
Chemical Name or Material Polyvinyl alcohol, Medium molecular weight, Hydrolyzed
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only