missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polysulfone resin, pellets, approx. M.W. 60,000, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 178910050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Polysulfone resin | |
| 25135-51-7 | |
| MFCD00134387 | |
| Solubility in water: insoluble. | |
| ZUZZUJNBGCVPLQ-UHFFFAOYSA-N | |
| 4-[2-(4-hydroxyphenyl)propan-2-yl]phenol;4-(4-hydroxyphenyl)sulfonylphenol | |
| 16211415 | |
| 1% max. | |
| 1.633 |
| nominal M.W. 75000(GPC) | |
| C27H26O6S | |
| 1.2400g/mL | |
| 5g | |
| CC(C)(C1=CC=C(C=C1)O)C2=CC=C(C=C2)O.C1=CC(=CC=C1O)S(=O)(=O)C2=CC=C(C=C2)O | |
| 478.559 | |
| Authentic | |
| Glass Bottle | |
| 1.24 |
RUO â Research Use Only