Learn More
Applied Biosystems™ Polystyrene Synthesis Column, ABI LV40™, dC(Bz)
Catalog No. 401743
Encompass
Change view
Click to view available options
Quantity:
1 Column
Catalog No. Quantity
401743 1 Column
1 options
Catalog No. 401743 Supplier Applied Biosystems™ Supplier No. 401743
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The dC(Bz) ABI LV40™ columns employ the advantages of polystyrene and a low volume column design to deliver high-quality oligonucleotides, while saving time and money.
Save Time and MoneyCompared to conventional columns, the ABI LV40™ columns reduce cycle time by about 10% and the cost of synthesis by about 25%. These columns are fully compatible with the Applied Biosystems 3400 instruments, ABI 391, ABI 392, ABI 394, and earlier instruments.
  • Proprietary design and manufacturing methods ensure consistency and high quality.
  • Each column is tested to verify accuracy and precision of fill, then pressure-tested to verify proper assembly and to guarantee each column is leak-free.
  • Hydrophobic polystryrene columns sustain anhydrous environment to promote efficient coupling while minimizing nonspecific binding sites, yielding an efficient synthesis.
  • The 1,000 angstrom pore size of the polystyrene support allows synthesis of oligonucleotides of varying length.
Specifications
Packaging Type Shipped at Room Temperature
Quantity 1 Column
For Research Use Only. Not for use in diagnostic procedures.