Learn More
Fisherbrand™ Polystyrene Storage Bottles
Reduce the risk of cross contamination with sterile, disposable polystyrene media bottles. Use with Bottle-Top Filters or as replacement/extra receptacles for Filter Units. Excellent as single-use storage containers.
Supplier: Fisherbrand™ FB12566515
Comprend: Individually packaged sterile screw top style bottle cap
Description
- Rounded with molded in hand grips
- Polystyrene
- Molded in graduations
- Sterile
Certifications
Non-pyrogenic per USP Standard Lot-to-Lotnon-cytotoxic Lot-to-Lot sterility
Spécifications
| Rounded with molded in hand grips | |
| Sterile | |
| 500 mL |
| Polystyrene | |
| Receiver Flask |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.